Aminoacetone hydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 1-Aminopropan-2-one hydrochloride

A186558-1G A186558-250MG A186558-25G A186558-5G A186558-100G
ALA186558-1GEA 56.91 USD
ALA186558-1G ALA186558250MG ALA186558-25G ALA186558-5G ALA186558-100G
Aminoacetone hydrochloride ≥97%
Aminoacetone hydrochloride
Formula: C₃H₈ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD00056125
CAS Number: 7737-17-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=318591da04d91c2fde49c1627ce2ab5db6820fe6&vid=318591da04d91c2fde49c1627ce2ab5db6820fe6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR