5-Amino-2-(4-aminophenyl)benzimidazole ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A186464-1G A186464-25G A186464-100G
ALA186464-1GEA 36.96 USD
ALA186464-1G ALA186464-25G ALA186464-100G
5-Amino-2-(4-aminophenyl)benzimidazole ≥98%
5-Amino-2-(4-aminophenyl)benzimidazole
Formula: C₁₃H₁₂N₄
Melting Pt: 236 °C
Storage Temperature: Ambient
MDL Number: MFCD00451475
CAS Number: 7621-86-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c6d9cce49da319e2993e473a78b727b160f8b90&vid=3c6d9cce49da319e2993e473a78b727b160f8b90&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR