5-Amino-8-chloroquinoline ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A186442-1G A186442-5G
ALA186442-1GEA 147.16 USD
ALA186442-1G ALA186442-5G
5-Amino-8-chloroquinoline ≥98%
5-Amino-8-chloroquinoline
Formula: C₉H₇ClN₂
MW: 178.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09802113
CAS Number: 75793-58-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=249ed641ebf8f9a559f8a6298d751204f616a5c1&vid=249ed641ebf8f9a559f8a6298d751204f616a5c1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR