2-Acetamido-3-methylpyridine ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A186388-1G
A186388-250MG
A186388-25G
A186388-5G
ALA186388-1GEA
63.56
USD
ALA186388-1G
ALA186388250MG
ALA186388-25G
ALA186388-5G
2-Acetamido-3-methylpyridine ≥98%
2-Acetamido-3-methylpyridine
Formula:
C₈H₁₀N₂O Boiling Pt: 158 °C Melting Pt: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00495347 CAS Number: 7463-30-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...