6-Acetylquinoline ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A186290-5G A186290-100MG A186290-1G A186290-250MG
ALA186290-5GEA 655.41 USD
ALA186290-5G ALA186290100MG ALA186290-1G ALA186290250MG
6-Acetylquinoline ≥97%
6-Acetylquinoline
Formula: C₁₁H₉NO
MW: 171.195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047614
CAS Number: 73013-68-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eecfb62a0ebadc27f3f57582a31bfcee8b2bbaa0&vid=eecfb62a0ebadc27f3f57582a31bfcee8b2bbaa0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR