3-Amino-3-(3-chlorophenyl)propionic acid ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A185995-1G A185995-250MG A185995-25G A185995-5G
ALA185995-1GEA 117.71 USD
ALA185995-1G ALA185995250MG ALA185995-25G ALA185995-5G
3-Amino-3-(3-chlorophenyl)propionic acid ≥95%
3-Amino-3-(3-chlorophenyl)propionic acid
Formula: C₉H₁₀ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00463160
CAS Number: 68208-21-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd9b0d4481be2c9be3a18af413bbdd2912fd6f4d&vid=cd9b0d4481be2c9be3a18af413bbdd2912fd6f4d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR