3-Amino-3-(3-methoxyphenyl)propionic acid ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 3-Amino-3-(3-methoxyphenyl)propanoic acid

A185994-1G A185994-250MG A185994-5G
ALA185994-1GEA 68.31 USD
ALA185994-1G ALA185994250MG ALA185994-5G
3-Amino-3-(3-methoxyphenyl)propionic acid ≥95%
3-Amino-3-(3-methoxyphenyl)propionic acid
Formula: C₁₀H₁₃NO₃
MW: 195.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00462569
CAS Number: 68208-19-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e8316a4c55358ac591fe67bd66c550d23a6a654&vid=3e8316a4c55358ac591fe67bd66c550d23a6a654&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR