5-Amino-2-methoxy-4-picoline ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 6-Methoxy-4-methylpyridin-3-amine, 5-Amino-2-methoxy-4-methylpyridine

A185897-1G A185897-25G A185897-5G
ALA185897-1GEA 15.11 USD
ALA185897-1G ALA185897-25G ALA185897-5G
5-Amino-2-methoxy-4-picoline ≥98%
5-Amino-2-methoxy-4-picoline
Formula: C₇H₁₀N₂O
MW: 138.1700 g/mol
Melting Pt: 157-161°C
Flash Pt: 124°C
Storage Temperature: Refrigerator
MDL Number: MFCD08277274
CAS Number: 6635-91-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddf9efc21df149758ffacf8c5cb84eea180cb388&vid=ddf9efc21df149758ffacf8c5cb84eea180cb388&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR