2-Amino-5-(trifluoromethyl)benzonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 4-Amino-3-cyanobenzotrifluoride, 2-Amino-5-trifluoromethylbenzonitrile

A185826-25G A185826-100G A185826-1G
ALA185826-25GEA 392.26 USD
ALA185826-25G ALA185826-100G ALA185826-1G
2-Amino-5-(trifluoromethyl)benzonitrile ≥98%
2-Amino-5-(trifluoromethyl)benzonitrile
Formula: C₈H₅F₃N₂
MW: 186.1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03407465
CAS Number: 6526-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR