3-Amino-4-(methylamino)benzonitrile ≥96%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A185803-1G A185803-250MG A185803-25G A185803-5G
ALA185803-1GEA 60.71 USD
ALA185803-1G ALA185803250MG ALA185803-25G ALA185803-5G
3-Amino-4-(methylamino)benzonitrile ≥96%
3-Amino-4-(methylamino)benzonitrile
Formula: C₈H₉N₃
MW: 147.177 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03407328
CAS Number: 64910-46-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd6f99c0f4f91b90ea3289af6ef4af8a7552d510&vid=cd6f99c0f4f91b90ea3289af6ef4af8a7552d510&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR