4-(Aminosulphonyl)benzamide ≥95%
Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning
|
Synonyms:
4-Sulfamoylbenzamide
A185681-1G
A185681-250MG
A185681-5G
ALA185681-1GEA
103.46
USD
ALA185681-1G
ALA185681250MG
ALA185681-5G
4-(Aminosulphonyl)benzamide ≥95%
4-(Aminosulphonyl)benzamide
Formula:
C₇H₈N₂O₃S MW: 200.2 g/mol Storage Temperature: Ambient |
CAS Number:
6306-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...