1-(2-Aminoethyl)-2-imidazolidone ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 1-(2-Aminoethyl)imidazolidin-2-one

A185652-1G A185652-25G A185652-5G
ALA185652-1GEA 23.66 USD
ALA185652-1G ALA185652-25G ALA185652-5G
1-(2-Aminoethyl)-2-imidazolidone ≥97%
1-(2-Aminoethyl)-2-imidazolidone
Formula: C₅H₁₁N₃O
MW: 129.16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00086348
CAS Number: 6281-42-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11264d433db327d1e4cfdb6e49a2166923b0bf6a&vid=11264d433db327d1e4cfdb6e49a2166923b0bf6a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR