3-Amino-N,N-dimethylbenzenesulfonamide ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A185646-5G
A185646-1G
A185646-250MG
ALA185646-5GEA
245.96
USD
ALA185646-5G
ALA185646-1G
ALA185646250MG
3-Amino-N,N-dimethylbenzenesulfonamide ≥98%
3-Amino-N,N-dimethylbenzenesulphonamide
Formula:
C₈H₁₂N₂O₂S Storage Temperature: Ambient |
MDL Number:
MFCD00602451 CAS Number: 6274-18-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...