2-Aminomalonamide ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A185601-100G A185601-1G A185601-25G A185601-5G
ALA185601-100GEA 54.06 USD
ALA185601-100G ALA185601-1G ALA185601-25G ALA185601-5G
2-Aminomalonamide ≥95%
2-Aminomalonamide
Formula: C₃H₇N₃O₂
MW: 117.1100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00015932
CAS Number: 62009-47-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60cb87b91f2b92c588efeab2bb48f46eb0cf8239&vid=60cb87b91f2b92c588efeab2bb48f46eb0cf8239&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR