3-Amino-4-methylbenzonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A185526-100G A185526-10G A185526-25G A185526-5G
ALA185526-100GEA 108.21 USD
ALA185526-100G ALA185526-10G ALA185526-25G ALA185526-5G
3-Amino-4-methylbenzonitrile ≥98%
3-Amino-4-methylbenzonitrile
Formula: C₈H₈N₂
MDL Number: MFCD04039781
CAS Number: 60710-80-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6adf9d5448a193f2f9d428b53814305abe196aef&vid=6adf9d5448a193f2f9d428b53814305abe196aef&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR