1-Acetylisonipecotoyl chloride 95

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 1-Acetylpiperidine-4-carbonyl chloride

A185426-1G A185426-25G A185426-5G
ALA185426-1GEA 68.31 USD
ALA185426-1G ALA185426-25G ALA185426-5G
1-Acetylisonipecotoyl chloride 95
1-Acetylisonipecotoyl chloride
Formula: C₈H₁₂ClNO₂
MW: 189.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02094017
CAS Number: 59084-16-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fdd020599efafd6bb363d547cf0d7566e70ce050&vid=fdd020599efafd6bb363d547cf0d7566e70ce050&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR