2-Amino-6-(trifluoromethyl)benzonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 2-Amino-6-trifluoromethylbenzonitrile

A185374-1G A185374-25G A185374-5G
ALA185374-1GEA 59.76 USD
ALA185374-1G ALA185374-25G ALA185374-5G
2-Amino-6-(trifluoromethyl)benzonitrile ≥98%
2-Amino-6-(trifluoromethyl)benzonitrile
Formula: C₈H₅F₃N₂
MW: 186.1339 g/mol
MDL Number: MFCD08741381
CAS Number: 58458-11-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e927afa657f7a4b05a93528f33925fa7ba410da&vid=5e927afa657f7a4b05a93528f33925fa7ba410da&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR