2-Bromo-3-(trifluoromethyl)aniline ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A185373-5G A185373-1G
ALA185373-5GEA 76.86 USD
ALA185373-5G ALA185373-1G
2-Bromo-3-(trifluoromethyl)aniline ≥98%
2-Bromo-3-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
MW: 240.02
Storage Temperature: Ambient
MDL Number: MFCD01656033
CAS Number: 58458-10-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6012fdd0b3b399effdfd793775cac3722681dbbe&vid=6012fdd0b3b399effdfd793775cac3722681dbbe&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR