7-Aminoquinoline ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A185350-1G A185350-5G A185350-250MG
ALA185350-1GEA 93.96 USD
ALA185350-1G ALA185350-5G ALA185350-250MG
7-Aminoquinoline ≥97%
7-Aminoquinoline
Formula: C₉H₈N₂
Melting Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD01646243
CAS Number: 580-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3c784aa533cc510a03e49a87112ce2f3fcfd976&vid=a3c784aa533cc510a03e49a87112ce2f3fcfd976&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR