5-Amino-2-bromopyrimidine ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A185256-1G A185256-250MG A185256-50MG A185256-5G
ALA185256-1GEA 93.01 USD
ALA185256-1G ALA185256250MG ALA185256-50MG ALA185256-5G
5-Amino-2-bromopyrimidine ≥95%
5-Amino-2-bromopyrimidine
Formula: C₄H₄BrN₃
Storage Temperature: Refrigerator
MDL Number: MFCD07778608
CAS Number: 56621-91-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8de4cdfe842d87eaff62160d04d8e51cf52415b2&vid=8de4cdfe842d87eaff62160d04d8e51cf52415b2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR