6-Aminopyridin-3-ol ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 6-Aminopyridin-3-ol, 2-Amino-5-hydroxypyridine

A185200-100MG A185200-1G A185200-250MG A185200-5G A185200-50MG
ALA185200100MGEA 22.71 USD
ALA185200100MG ALA185200-1G ALA185200250MG ALA185200-5G ALA185200-50MG
6-Aminopyridin-3-ol ≥95%
6-Aminopyridin-3-ol
Formula: C₅H₆N₂O
MW: 110.1100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03541056
CAS Number: 55717-46-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b58c8ea4651d08bd18dc6c8d31cd6c9baf8d0f67&vid=b58c8ea4651d08bd18dc6c8d31cd6c9baf8d0f67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR