2-Amino-6-methylpyrazine ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 6-Methylpyrazin-2-amine

A185169-1G A185169-25G A185169-5G
ALA185169-1GEA 85.41 USD
ALA185169-1G ALA185169-25G ALA185169-5G
2-Amino-6-methylpyrazine ≥97%
2-Amino-6-methylpyrazine
Formula: C₅H₇N₃
MW: 109.13 g/mol
Melting Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD08235193
CAS Number: 5521-56-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a437eda40c522ed9ef462534d30d107828c27b4&vid=3a437eda40c522ed9ef462534d30d107828c27b4&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR