2-Amino-5-bromobenzenesulphonamide 96

Supplier: ALADDIN SCIENTIFIC
Warning

A185142-1G A185142-5G
ALA185142-1GEA 162.36 USD
ALA185142-1G ALA185142-5G
2-Amino-5-bromobenzenesulphonamide 96
2-Amino-5-bromobenzenesulphonamide
Formula: C₆H₇BrN₂O₂S
MW: 251.1
Melting Pt: 170-176 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03426835
CAS Number: 54734-84-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca35330db5efc7264010324829794e96f3e287a0&vid=ca35330db5efc7264010324829794e96f3e287a0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR