2-Amino-N,N-dimethylbenzenesulfonamide ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A185119-100MG A185119-1G A185119-250MG A185119-5G A185119-25G
ALA185119100MGEA 17.96 USD
ALA185119100MG ALA185119-1G ALA185119250MG ALA185119-5G ALA185119-25G
2-Amino-N,N-dimethylbenzenesulfonamide ≥98%
2-Amino-N,N-dimethylbenzenesulphonamide
Formula: C₈H₁₂N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD04035363
CAS Number: 54468-86-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bf27009013a59529a8416ea78854979bb0c291f&vid=1bf27009013a59529a8416ea78854979bb0c291f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR