5-Amino-1-methyl-1H-pyrazole-4-carbonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A185036-25G A185036-5G
ALA185036-25GEA 149.06 USD
ALA185036-25G ALA185036-5G
5-Amino-1-methyl-1H-pyrazole-4-carbonitrile ≥98%
5-Amino-1-methyl-1H-pyrazole-4-carbonitrile
Formula: C₅H₆N₄
MDL Number: MFCD00114569
CAS Number: 5334-41-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da6f309830f55e026b410d393d75804955659190&vid=da6f309830f55e026b410d393d75804955659190&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR