4-Amino-2-fluorobenzonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A185034-1G A185034-100G
ALA185034-1GEA 15.11 USD
ALA185034-1G ALA185034-100G
4-Amino-2-fluorobenzonitrile ≥98%
4-Amino-2-fluorobenzonitrile
Formula: C₇H₅FN₂
MW: 136.1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368709
CAS Number: 53312-80-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2fad67ca92226e0120c99f67baeb230a3703bdc&vid=c2fad67ca92226e0120c99f67baeb230a3703bdc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR