3-Amino-5-chlorobenzonitrile ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
A185033-1G
A185033-250MG
A185033-5G
ALA185033-1GEA
36.01
USD
ALA185033-1G
ALA185033250MG
ALA185033-5G
3-Amino-5-chlorobenzonitrile ≥98%
3-Amino-5-chlorobenzonitrile
Formula:
C₇H₅ClN₂ MW: 152.58 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06797224 CAS Number: 53312-78-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...