5-Amino-3-bromo-2-methoxypyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 5-Bromo-6-methoxypyridin-3-amine

A185025-100G A185025-1G A185025-25G A185025-5G
ALA185025-100GEA 302.01 USD
ALA185025-100G ALA185025-1G ALA185025-25G ALA185025-5G
5-Amino-3-bromo-2-methoxypyridine ≥98%
5-Amino-3-bromo-2-methoxypyridine
Formula: C₆H₇BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD06659518
CAS Number: 53242-18-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78f394ecf0b4a0b30ea84738f7e4a41e01acb292&vid=78f394ecf0b4a0b30ea84738f7e4a41e01acb292&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR