Methyl 5-(4-aminophenyl)furan-2-carboxylate ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 5-(4-Aminophenyl)furan-2-carboxylic acid methyl ester

A185001-1G A185001-250MG A185001-5G
ALA185001-1GEA 91.11 USD
ALA185001-1G ALA185001250MG ALA185001-5G
Methyl 5-(4-aminophenyl)furan-2-carboxylate ≥95%
Methyl 5-(4-aminophenyl)furan-2-carboxylate
Formula: C₁₂H₁₁NO₃
MW: 217.22 g/mol
Melting Pt: 122-126 °C
MDL Number: MFCD06797396
CAS Number: 52939-06-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a4ff0fd83ca8cc6643f76a47c6731852944de5e&vid=7a4ff0fd83ca8cc6643f76a47c6731852944de5e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR