1-[(Ammoniooxy)methyl]-4-fluorobenzene chloride ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A184897-10G A184897-1G A184897-25G A184897-5G
ALA184897-10GEA 173.76 USD
ALA184897-10G ALA184897-1G ALA184897-25G ALA184897-5G
1-[(Ammoniooxy)methyl]-4-fluorobenzene chloride ≥98%
1-[(Ammoniooxy)methyl]-4-fluorobenzene chloride
Formula: C₇H₉ClFNO
Storage Temperature: Ambient
MDL Number: MFCD01114581
CAS Number: 51572-89-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aed74ae2b7486adc91a2dfc5fc2614395382ec91&vid=aed74ae2b7486adc91a2dfc5fc2614395382ec91&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR