2-Amino-2-methylpropanenitrile hydrochloride ≥90%

Supplier: ALADDIN SCIENTIFIC
Danger

A184859-25G A184859-5G
ALA184859-25GEA 18.91 USD
ALA184859-25G ALA184859-5G
2-Amino-2-methylpropanenitrile hydrochloride ≥90%
2-Amino-2-methylpropanenitrile hydrochloride
Formula: C₄H₈N₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD08456816
CAS Number: 50846-36-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c7d1c0d3fa57bfa21f05ca768d063fd8451b548&vid=4c7d1c0d3fa57bfa21f05ca768d063fd8451b548&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR