4-Fluoro-3-methoxybenzylamine ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 5-Aminomethyl-2-fluoroanisole, 5-(Aminomethyl)-2-fluoroanisole

A184856-1G A184856-250MG A184856-5G
ALA184856-1GEA 97.76 USD
ALA184856-1G ALA184856250MG ALA184856-5G
4-Fluoro-3-methoxybenzylamine ≥98%
4-Fluoro-3-methoxybenzylamine
Formula: C₈H₁₀FNO
MW: 155.1700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04116362
CAS Number: 508177-67-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ccffbfaca8706e6eff195f847649d8ade6a1242c&vid=ccffbfaca8706e6eff195f847649d8ade6a1242c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR