1-(2-Amino-3,4-dimethoxyphenyl)ethanone ≥95%

Supplier: ALADDIN SCIENTIFIC

A184774-1G A184774-250MG A184774-50MG
ALA184774-1GEA 289.66 USD
ALA184774-1G ALA184774250MG ALA184774-50MG
1-(2-Amino-3,4-dimethoxyphenyl)ethanone ≥95%
1-(2-Amino-3,4-dimethoxyphenyl)ethanone
Formula: C₁₀H₁₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD11040634
CAS Number: 49701-79-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1ab3ed5d2b5f6d00b8b2216e6a8adcc75ef2ec3&vid=b1ab3ed5d2b5f6d00b8b2216e6a8adcc75ef2ec3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR