3-Amino-5-bromobenzonitrile ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 5-Amino-3-bromobenzonitrile

A184771-10G A184771-1G A184771-250MG A184771-5G
ALA184771-10GEA 283.01 USD
ALA184771-10G ALA184771-1G ALA184771250MG ALA184771-5G
3-Amino-5-bromobenzonitrile ≥97%
3-Amino-5-bromobenzonitrile
Formula: C₇H₅BrN₂
MW: 197.0300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11109874
CAS Number: 49674-16-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12d281682afbc0058c3df347df43df90816afc32&vid=12d281682afbc0058c3df347df43df90816afc32&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR