2-Amino-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid hydrochloride ≥95%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 2-Amino-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid hydrochloride, 3-(2-Oxo-1,2-dihydro-4-quinolinyl)-DL-alanine hydrochloride

A184740-100G A184740-1G A184740-250MG A184740-25G A184740-5G
ALA184740-100GEA 1970.21 USD
ALA184740-100G ALA184740-1G ALA184740250MG ALA184740-25G ALA184740-5G
2-Amino-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid hydrochloride ≥95%
2-Amino-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid hydrochloride
Formula: C₁₂H₁₃ClN₂O₃
MW: 268.70 g/mol
Melting Pt: 225°C
Storage Temperature: Refrigerator
MDL Number: MFCD07787426
CAS Number: 4876-14-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d404c05cb23c13e3d47f20da096f0d41140716ae&vid=d404c05cb23c13e3d47f20da096f0d41140716ae&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR