3-Amino-6-bromopyrazine-2-carboxylic acid ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A184736-10G A184736-1G A184736-25G A184736-5G
ALA184736-10GEA 457.81 USD
ALA184736-10G ALA184736-1G ALA184736-25G ALA184736-5G
3-Amino-6-bromopyrazine-2-carboxylic acid ≥98%
3-Amino-6-bromopyrazine-2-carboxylic acid

3-Amino-6-bromopyrazine-2-carboxylic acid used to prepare 3-bromo-5-phenylpyrazine-2-amine. It serves as an intermediate in organic synthesis and pharmaceuticals.

Formula: C₅H₄BrN₃O₂
MW: 218.01 g/mol
Melting Pt: 178-183°C
Storage Temperature: Ambient
MDL Number: MFCD08275681
CAS Number: 486424-37-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67919708ec1bbdb8b78c328a8ad2a5a36d8ca0b1&vid=67919708ec1bbdb8b78c328a8ad2a5a36d8ca0b1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR