2-Amino-5-phenyl-thiophene-3-carboxylic acid amide ≥96%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 2-Amino-5-phenyl-3-thiophenecarboxamide

A184723-1G A184723-25G A184723-5G
ALA184723-1GEA 84.46 USD
ALA184723-1G ALA184723-25G ALA184723-5G
2-Amino-5-phenyl-thiophene-3-carboxylic acid amide ≥96%
2-Amino-5-phenyl-thiophene-3-carboxylic acid amide

Product Application:Product Uses To be used only for scientific research and development. Not for use in humans or animals.

Formula: C₁₁H₁₀N₂OS
MW: 218.3 g/mol
Storage Temperature: Refrigerator
CAS Number: 4815-35-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbe3a70e952a601f5b44fb7aad2446cf40a21a19&vid=dbe3a70e952a601f5b44fb7aad2446cf40a21a19&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR