6-Amino-5-bromonicotinonitrile ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A184685-100MG
A184685-10G
A184685-1G
A184685-250MG
A184685-5G
ALA184685100MGEA
17.96
USD
ALA184685100MG
ALA184685-10G
ALA184685-1G
ALA184685250MG
ALA184685-5G
6-Amino-5-bromonicotinonitrile ≥98%
6-Amino-5-bromonicotinonitrile
Formula:
C₆H₄BrN₃ MW: 198.0200 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD02102412 CAS Number: 477871-32-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...