2-Amino-4-methylthiophene-3-carbonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Amino-3-cyano-4-methylthiophene

A184640-1G A184640-25G A184640-5G
ALA184640-1GEA 235.51 USD
ALA184640-1G ALA184640-25G ALA184640-5G
2-Amino-4-methylthiophene-3-carbonitrile ≥98%
2-Amino-4-methylthiophene-3-carbonitrile
Formula: C₆H₆N₂S
MW: 138.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00277568
CAS Number: 4623-55-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22fb926e97170157a5966b91620cf06f0718662f&vid=22fb926e97170157a5966b91620cf06f0718662f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR