2-Amino-4'-fluoroacetophenone hydrochloride ≥96%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Amino-1-(4-fluorophenyl)ethanone hydrochloride, 2-Amino-4'-fluoroacetophenone Hydrochloride, 4-Fluorophenacylamine hydrochloride

A184615-100G A184615-1G A184615-250MG A184615-25G A184615-5G
ALA184615-100GEA 1088.61 USD
ALA184615-100G ALA184615-1G ALA184615250MG ALA184615-25G ALA184615-5G
2-Amino-4'-fluoroacetophenone hydrochloride ≥96%
2-Amino-4'-fluoroacetophenone hydrochloride
Formula: C₈H₉ClFNO
MW: 189.6146 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00193059
CAS Number: 456-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42a87226e799a8b6f1554958f1d11736ab47c0e6&vid=42a87226e799a8b6f1554958f1d11736ab47c0e6&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR