Isonicotinamidine xhydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Carbamimidoylpyridine xhydrochloride, Pyridine-4-carboxamidine xhydrochloride, 4-Pyridinecarboximidamide Hydrochloride, 4-Amidinopyridine xhydrochloride, Pyridine-4-carboximidamide xhydrochloride, Isonicotinimidamide xhydrochloride

A184477-100G A184477-25G A184477-5G
ALA184477-100GEA 596.51 USD
ALA184477-100G ALA184477-25G ALA184477-5G
Isonicotinamidine xhydrochloride ≥97%
Isonicotinamidine hydrochloride
Formula: C₆H₇N₃·xClH
MW: 157.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040385
CAS Number: 42518-06-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8508333170807203ae24acc2f3bcdd497d407eb1&vid=8508333170807203ae24acc2f3bcdd497d407eb1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR