2-Amino-5-chloropyridin-3-ol ≥95%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
2-Amino-5-chloro-3-hydroxypyridine, 2-Amino-3-hydroxy-5-chloropyridine
A184404-1G
A184404-250MG
A184404-25G
A184404-5G
ALA184404-1GEA
21.76
USD
ALA184404-1G
ALA184404250MG
ALA184404-25G
ALA184404-5G
2-Amino-5-chloropyridin-3-ol ≥95%
2-Amino-5-chloropyridin-3-ol
Formula:
C₅H₅ClN₂O MW: 144.5590 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14706179 CAS Number: 40966-87-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...