4-Amino-1-(methylsulfonyl)piperidine ≥98%

Supplier: Aladdin Scientific
Warning

A184342-1G A184342-250MG
ALA184342-1GEA 15.11 USD
ALA184342-1G ALA184342250MG
4-Amino-1-(methylsulfonyl)piperidine ≥98%
4-Amino-1-(methylsulfonyl)piperidine
Formula: C₆H₁₄N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD06805736
CAS Number: 402927-97-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a47f5244f92f410b56a9040bd398afbf7208ba9&vid=1a47f5244f92f410b56a9040bd398afbf7208ba9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR