2-Amino-4-thiazolecarboxylic acid ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A184340-100G A184340-25G A184340-5G
ALA184340-100GEA 195.9 USD
ALA184340-100G ALA184340-25G ALA184340-5G
2-Amino-4-thiazolecarboxylic acid ≥98%
2-Amino-4-thiazolecarboxylic acid
Formula: C₄H₄N₂O₂S
MDL Number: MFCD00859429
CAS Number: 40283-41-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36bee8e51b965e5e88c5e01207d231b98007e341&vid=36bee8e51b965e5e88c5e01207d231b98007e341&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR