Ethyl 1-(aminomethyl)cyclopropanecarboxylate ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1-(Aminomethyl)-cyclopropanecarboxylic acid ethyl ester

A184318-100MG A184318-1G A184318-250MG A184318-5G
ALA184318100MGEA 26.51 USD
ALA184318100MG ALA184318-1G ALA184318250MG ALA184318-5G
Ethyl 1-(aminomethyl)cyclopropanecarboxylate ≥95%
Ethyl 1-(aminomethyl)cyclopropanecarboxylate
Formula: C₇H₁₃NO₂
MW: 143.18 g/mol
Storage Temperature: Freezer
MDL Number: MFCD06212652
CAS Number: 400840-94-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a24ff0fed5a04ea9a0ecf5f25340cf5db61b64b9&vid=a24ff0fed5a04ea9a0ecf5f25340cf5db61b64b9&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR