4-Amino-α,α,α-trifluoro-m-toluic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Amino-3-(trifluoromethyl)benzoic acid

A184315-1G A184315-5G
ALA184315-1GEA 164.26 USD
ALA184315-1G ALA184315-5G
4-Amino-α,α,α-trifluoro-m-toluic acid ≥98%
4-Amino-α,α,α-trifluoro-m-toluic acid
Formula: C₈H₆F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD03407960
CAS Number: 400-76-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc32758eb12b1eeb94f342b5d4abc7129a94363d&vid=fc32758eb12b1eeb94f342b5d4abc7129a94363d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR