4-Aminomethylbenzyl alcohol ≥96%

Supplier: ALADDIN SCIENTIFIC
Danger

A184295-1G A184295-250MG A184295-5G
ALA184295-1GEA 59.76 USD
ALA184295-1G ALA184295250MG ALA184295-5G
4-Aminomethylbenzyl alcohol ≥96%
4-Aminomethylbenzyl alcohol
Formula: C₈H₁₁NO
Storage Temperature: Refrigerator
MDL Number: MFCD06213766
CAS Number: 39895-56-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cdb8d78cfabfb0fe4907901c9334b1f818afec1d&vid=cdb8d78cfabfb0fe4907901c9334b1f818afec1d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR