5-Phenyl-1,3-thiazol-2-amine ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
5-Phenylthiazol-2-amine, 2-Amino-5-phenylthiazole, 5-Phenyl-1,3-thiazol-2-amine, 5-Phenyl-2-thiazolamine
A184243-1G
A184243-250MG
A184243-25G
A184243-5G
ALA184243-1GEA
79.71
USD
ALA184243-1G
ALA184243250MG
ALA184243-25G
ALA184243-5G
5-Phenyl-1,3-thiazol-2-amine ≥98%
5-Phenyl-1,3-thiazol-2-amine
Formula:
C₉H₈N₂S Melting Pt: 206°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00022451 CAS Number: 39136-63-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...