4-(2-Aminoethyl)tetrahydropyran hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2-(Tetrahydro-2H-pyran-4-yl)ethanamine hydrochloride, 4-(2-Aminoethyl)tetrahydropyran, HCl

A184236-100MG A184236-1G A184236-250MG
ALA184236100MGEA 105.36 USD
ALA184236100MG ALA184236-1G ALA184236250MG
4-(2-Aminoethyl)tetrahydropyran hydrochloride ≥98%
4-(2-Aminoethyl)tetrahydropyran hydrochloride
Formula: C₇H₁₆ClNO
MW: 165.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411927
CAS Number: 389621-77-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43d2f6aecea19d88a2b9c650ad3c0005d90bf86c&vid=43d2f6aecea19d88a2b9c650ad3c0005d90bf86c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR