5-Bromo-3-chloropyridin-2-amine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Amino-5-bromo-3-chloropyridine, 5-Bromo-3-chloro-2-pyridinamine

A184168-100G A184168-1G A184168-250MG A184168-25G A184168-5G
ALA184168-100GEA 626.91 USD
ALA184168-100G ALA184168-1G ALA184168250MG ALA184168-25G ALA184168-5G
5-Bromo-3-chloropyridin-2-amine ≥98%
5-Bromo-3-chloropyridin-2-amine
Formula: C₅H₄BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD11110443
CAS Number: 38185-55-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=249d3496b7291be26cff6ae9082a82bbc174f273&vid=249d3496b7291be26cff6ae9082a82bbc174f273&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR